Distillation Receivers, Sampling, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This receiver is specially designed to receiver draw samples during distillation, or to return the condensed vapor to the flask
Fabricated from borosilicate glass.
Customers Who Bought This Also Bought