Gelex Secondary Standards Kit, 2100 Series Portable Turbidimeters, Hach
# de catalogue CA97027-030
Fournisseur: Hach
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Gelex Secondary Standards Kit
- Size:3 sealed vials
- Cat. no.:CA97027-030
- For Use With:2100 Series Portable Turbidimeters
- Supplier No.:2464105
Spécifications
A propos de cet article
Gelex Secondary Standards Kit, for 2100P and 2100Q Series Portable Turbidimeters (includes 1 sealed vial each, 0-10 NTU, 10-100 NTU, 100-1000 NTU).
Ordering information: 3/pk.
Customers Who Bought This Also Bought