WorkWipes® New White Terry Towel in Bag, New Pig
Catalogue # 89073-932
Fournisseur: New Pig
Spécifications
- Cat. no.:89073-932
- Packaging:Compressed
- Weight:9.1 kg (20 lbs.)
- Supplier No.:WIP581
Spécifications
À propos de cet article
All of our new rags are screened to assure material quality and performance.
- Ideal for cleaning and wiping in foodservice and food prep jobs
Eco- and cost-friendly rags made from 100% terry cotton are soft and very absorbent.
Compression-packed bags take up half the space of loosely packed boxes.
Strict material and size requirements mean you get more usable, high quality rags in every package. No useless rag pieces.
Not scanned for metal. Not sorted for printing. Not washed.