WorkWipes® New Colored T-Shirt in Bag, New Pig
Catalog # 89073-922
Supplier: New Pig
Specifications
- Cat. no.:89073-922
- Packaging:Compressed
- Weight:11.3 kg (25 lbs.)
- Supplier No.:WIP559
Specifications
About this item
All of our new rags are screened to assure material quality and performance.
- Durable
- Low-lint material is ideal for staining
- Polishing and general wiping
Eco- and cost-friendly rags are made from factory mill ends of new cotton and cotton-blend T-shirt material. More consistent performance and nicer appearance than reclaimed rags.
Compression-packed bags take up half the space of loosely packed boxes.
Strict material and size requirements mean you get more usable, high quality rags in every package. No useless rag pieces.
Not scanned for metal. Sorted for printing. Not washed.
Customers Who Bought This Also Bought