WorkWipes® Reclaimed Denim in Bag, New Pig
# de catalogue 89073-928
Fournisseur: New Pig
Spécifications
- Cat. no.:89073-928
- Packaging:Compressed
- Weight:11.3 kg (25 lbs.)
- Supplier No.:WIP575
Spécifications
A propos de cet article
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Especially efficient at absorbing oily liquids
- Excellent choice for steel mills or other high-temperature work sites
Eco- and cost-friendly rags are made from pant legs only - no rivets or zippers to mar performance. Thick, heavy denim rags are ideal for heavy-duty wiping.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Ordering information: Scanned for metal. Not sorted for printing. Not washed.
Customers Who Bought This Also Bought