WorkWipes® Reclaimed Surgical Huck Towel in Bag, New Pig
# de catalogue 89073-926
Fournisseur: New Pig
Spécifications
- Cat. no.:89073-926
- Packaging:Compressed
- Weight:9.1 kg (20 lbs.)
- Supplier No.:WIP570
Spécifications
A propos de cet article
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Colorfast, low-lint, sanitized, 100% cotton material is great for general cleaning and glass cleaning
Eco- and cost-friendly rags are made from reclaimed blue surgical huck towel material with hemmed edges.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Ordering information: Scanned for metal. Not sorted for printing. Washed.
Customers Who Bought This Also Bought