WorkWipes® Reclaimed Colored Flannel in Bag, New Pig
# de catalogue 89073-920
Fournisseur: New Pig
Spécifications
- Cat. no.:89073-920
- Packaging:Compressed
- Weight:11.3 kg (25 lbs.)
- Supplier No.:WIP557
Spécifications
A propos de cet article
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Durable, absorbent, soft, non-scratching material is ideal for wiping
Eco- and cost-friendly rags are made from recycled, post-consumer flannel shirts.
Compression-packed bags take up half the space of loosely packed boxes.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Scanned for metal. Not sorted for printing. Not washed.
Customers Who Bought This Also Bought