WorkWipes® Reclaimed White T-Shirt in Bag, New Pig
# de catalogue 89073-960
Fournisseur: New Pig
Spécifications
- Cat. no.:89073-960
- Packaging:Compressed
- Weight:11.3 kg (25 lbs.)
- Supplier No.:WIP561
Spécifications
A propos de cet article
Strict material and size requirements mean you get more usable, high quality rags in every package.
- Compression-packed bags take up half the space of loosely packed boxes
- Scanned for metal
- Sorted for printing
- Not washed
Eco- and cost-friendly rags are made from post-consumer recycled cotton and cotton blend T-shirts, so they're strong and absorbent. Durable, colorfast, low-lint material is ideal for staining, wiping, polishing and other general-use tasks. White color is ideal for use with solvents, which might cause colored rags to bleed.
No useless rag pieces. All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance.
Customers Who Bought This Also Bought