2-Ply Center-Pull Washroom Hand Towels, New Pig
Catalog # 89205-836
Supplier: 2 D 2 S.A. DE C.V. MX
Specifications
- Cat. no.:89205-836
- Packaging:600 wipers per roll, 6 rolls per case
- W×L:18.4 x 27.9 cm (7.25 x 11")
- Supplier No.:WRT601
Specifications
About this item
Embossed towels are great for drying face and hands.
- Center-pull roll fits wall-mounted unit
- Dispenses one at a time to help prevent overuse
- Center-pull rolls for easy, sanitary dispensing
Great for use in dispensers near sinks and basins, in restrooms, washrooms and kitchens. Also useful as an economical light-duty industrial wiper.
Frequently Bought Together