WorkWipes® Series 40 Wipers, New Pig
Catalog # 89205-866
Supplier: New Pig
Specifications
- Color:White
- Cat. no.:89205-866
- Packaging:1/4-fold
- W×L:29 x 32.4 cm (11.4 x 12.75")
- Supplier No.:WIP640
Specifications
About this item
Good for wiping up grease, oils and small spills or for cleaning parts and tools.
- Lower cost per wiper for high-volume use
Clothlike construction is soft enough for wiping hands and face. Medium-weight bonded cellulose-binder is forced deep into the soft fibers for extra strength. Available ¹/₄ folded and poly wrapped for easy handling and loading.
Customers Who Bought This Also Bought