UV Keychain Laser Light Pointer, Sper Scientific
# de catalogue 97035-720
Fournisseur: Sper Scientific
Spécifications
- Description:UV keychain laser light pointer
- Cat. no.:97035-720
- W×D×H:83×13.5 mm (3.25×0.5")
- Supplier No.:330006
Spécifications
A propos de cet article
Press one end of the button to activate a bright 650nm red laser pointer. Press the other end to activate a high intensity white LED light for inspection and illumination. Press the same side again to activate a 400 nm ultra-violet light.
- beam is sharp and very bright
- sturdy construction
- long battery life allow for extended
Ideal as a teaching tool for science presentations, conferences, lectures, seminars, and planetariums.
Weight: 38 g (1.3 oz.)
Packaging: All applications are packed into a tiny pocket-sized device.
Customers Who Bought This Also Bought