Microscope Pen, Sper Scientific
Catalog # 97035-716
Supplier: Sper Scientific
Specifications
- Description:Microscope Pen
- Cat. no.:97035-716
- Supplier No.:330004
Specifications
About this item
Useful for on the spot inspections in printing, textile, art, jewelry, microelectronics, and other industries
- Also serves as an inexpensive and simple to use microscope for science education and other labs
- Equipped with powerful 50x optics
- Weighing only 20g (0.7oz.), the pen clips easily to a shirt pocket and can be carried anywhere
135x10 mm (53/8 x 3/8")
Ordering information: The microscope pen is supplied with a 5-year warranty.
Customers Who Bought This Also Bought