Distillation Receivers, Graduated, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This is an excellent all-purpose receiver for general distillation work
Body is graduated. Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system. Stopcocks are 4mm bore. Tubing connections are 10mm O.D. olive type. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought