Disposable Standard-Tip Serological Pipettes, Plastic, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Standard-Tip Serological Pipettes are irradiation sterilized and suitable for tissue culture work.
They are equipped with uniform pulled or molded tips; beveled or molded top ends with cotton plugging. Two rings near the top denote that the small amount remaining in tip after delivery must be blown out and added to main volume delivered. The 5 ml size features a two-piece design. Color-coded packaging makes choosing the correct pipette size quick and accurate. Each pipette is equipped with negative and reverse graduations.
Certifications: Designed from ASTM E 934. Calibrated 'To Deliver' to tip.
Ordering information: Multipacked 10 ml pipettes are available with either regular tip (1.5 mm dia.) or wide tip (3.5 mm dia.). All sizes available in either paper/plastic individual wraps overpacked in dispensing shelf packs, or in recloseable multipacks overpacked in shipping containers.
Customers Who Bought This Also Bought