Light Meter Lux, Sper Scientific
Fournisseur: Sper Scientific
Certificats
A propos de cet article
Measure visible light levels anywhere with fast, stable, and accurate response
Certified light meter 57800-250 includes a NIST traceable certificate of calibration.
Results are easily read on the extra-large 22mm (⅞") LCD display, which also indicates when the battery is low or the reading over range. The color and cosine corrected light sensor meets C.I.E. photopic spectrum. The unit also features an external zero adjustment, hold function, and a tripod back.
Ordering information: Supplied ready to use with a soft padded case and shoulder strap, instructions, detachable sensor with cover, and a 9V battery. 5-year warranty included.
Spécifications
- Catalog No:
- 57800-211
- 57800-250
- Weight:
- 280 g (9 oz.)
- 280 g (9 oz.)
- Dimensions:
- 200 x 68 x 30 mm (8 x 23/4 x 11/4")
- 200 x 68 x 30 mm (8 x 23/4 x 11/4")