H-Gly-Pro-Gly-NH2 · HCl
Catalog # H-9865.1000BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:141497-12-3
- Size:1 g
- Protein/peptide name:H-Gly-Pro-Gly-NH2 · HCl
- Sequence:GPG-NH2 was shown to inhibit the replication of HIV-1 via its metabolite α-hydroxyglycineamide. The tripeptide added to the antiviral effect of both zidovudine and ritonavir.
- Molecular weight:264.71
- Molecular formula:C₉H₁₆N₄O₃ · HCl
- Storage conditions:–20 ± 5 °C
- Cat. no.:H-9865.1000BA
- Supplier No.:H-9865.1000
Specifications
About this item
GPG-NH2 was shown to inhibit the replication of HIV-1 via its metabolite α-hydroxyglycineamide. The tripeptide added to the antiviral effect of both zidovudine and ritonavir.
Sequence: H-Gly-Pro-Gly-NH2 · HCl
CAS Number: 141497-12-3
Molecular Formula: C9H16N4O3 · HCl
Molecular Weight: 264.71
Customers Who Bought This Also Bought