Human Recombinant pTH-Related Protein (1-86)
Catalog # H-9815.0500BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Size:0.5 mg
- Protein/peptide name:pTH-Related Protein (1-86)
- Protein synonyms:Hypercalcemia of Malignancy Factor (1-86)
- MDL No.:MFCD00163973
- Storage conditions:–20 ± 5 °C
- Cat. no.:H-9815.0500BA
- Supplier No.:H-9815.0500
Specifications
About this item
pTHrP (1-86) increased bone formation in vitro.
Customers Who Bought This Also Bought