Mitogenic Pentapeptide
# de catalogue H-9460.0005BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:87173-03-3
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237176
- Protein synonyms:Pam3Cys-Ser-Ser-Asn-Ala-OH|Pam3CSSNA
- Protein/peptide name:Mitogenic Pentapeptide
- Molecular weight:1269.82
- Sequence:Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-Ser-Ser-Asn-Ala-OH
- Molecular formula:C₆₇H₁₂₄N₆O₁₄S
- Cat. no.:H-9460.0005BA
- Supplier No.:H-9460.0005
Spécifications
A propos de cet article
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
The palmitoylated pentapeptide represents a potent activator for monocytes/macrophages and B lymphocytes.
Customers Who Bought This Also Bought