Neuropeptide Y (22-36) Trifluoroacetate
# de catalogue H-9305.0005BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:119019-65-7
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD03453649
- Protein/peptide name:Neuropeptide Y (22-36) Trifluoroacetate
- Molecular weight:1903.22
- Sequence:H-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH₂
- Molecular formula:C₈₅H₁₃₉N₂₉O₂₁
- Cat. no.:H-9305.0005BA
- Supplier No.:H-9305.0005
Spécifications
A propos de cet article
This C-terminal fragment of NPY is a more potent inhibitor of calmodulin-stimulated phosphodiesterase than NPY itself. The inhibitory effect obviously depends on the basic amphiphilic helical nature of the fragment.
Customers Who Bought This Also Bought