Bovine (D-Trp¹²,Tyr³⁴)-pTH (7-34) Amide Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
See also the pTH2 receptor agonist TIP39 (H-4878).
The D-Trp¹² substitution leads to a competitive pTH antagonist with increased inhibitory properties in vitro.
Customers Who Bought This Also Bought