(Leu³¹,Pro³⁴)-Neuropeptide Y Trifluoroacetate, Bachem
Catalog # H-8575.1000BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:125580-28-1
- Species:Pig
- Size:1 mg
- Protein/peptide name:(Leu³¹,Pro³⁴)-Neuropeptide Y Trifluoroacetate
- MDL No.:MFCD00166234
- Sequence:H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Leu-Thr-Arg-Pro-Arg-Tyr-NH₂
- Molecular weight:4222.69
- Molecular formula:C₁₉₀H₂₈₆N₅₄O₅₆
- Storage conditions:–20 ± 5 °C
- Cat. no.:H-8575.1000BA
- Supplier No.:H-8575.1000
Specifications
About this item
This NPY analog is a highly potent and selective ligand for Y₁ receptors in different cell lines and tissues. It has therefore been used to detect the NPY/PYY receptor subtype distribution, e.g., in rat brain. The peptide is also active in vivo, showing an even higher potency than NPY in increasing blood pressure in anesthetized rats.
Customers Who Bought This Also Bought