(Tyr⁵,D-Trp⁶·⁸·⁹,Lys-NH₂¹⁰)-Neurokinin A (4-10)
Supplier: Bachem Americas
Certificates
About this item
MEN10376 is a potent antagonist with high affinity and selectivity for the NK-2 receptor in vitro and in vivo.
Frequently Bought Together
MEN10376 is a potent antagonist with high affinity and selectivity for the NK-2 receptor in vitro and in vivo.