Human;Mouse;Rat Adropin (34-76) Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Adropin is a secreted factor involved in energy homeostasis and lipid metabolism. It is encoded by the energy homeostasis associated gene (Enho) and is expressed in liver and brain. In diet-induced obesity (DIO) mice Adropin (34-76) attenuated hepatosteatosis and insulin reistance independently of adiposity or food intake. Additionally, adropin could be a regulator of endothelial function.
Customers Who Bought This Also Bought