About this item
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
GnRH antagonist used in the treatment of hormone-sensitive prostate cancer. CAS Number (cetrorelix acetate): 145672-81-7. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.