Bovine;Horse;Human;Rat Thymosin beta₄ Acetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Thymosin β₄ is a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide. Extracellular thymosin β₄ may contribute to physiological processes such as angiogenesis, wound healing, and regulation of inflammation. Additionally to its numerous functions thymosin β₄ might also be of therapeutic value in the setting of acute myocardial damage.
Customers Who Bought This Also Bought