H-Ala-Gly-Ala-OH, Bachem
# de catalogue H-1480.0001BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:37460-22-3
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00190743
- Protein/peptide name:H-Ala-Gly-Ala-OH
- Molecular weight:217.23
- Sequence:H-Ala-Gly-Ala-OH
- Molecular formula:C₈H₁₅N₃O₄
- Cat. no.:H-1480.0001BA
- Supplier No.:H-1480.0001
Spécifications
A propos de cet article
Customers Who Bought This Also Bought