Bovine;Human;Mouse Thrombospondin-1 (1016-1023)
# de catalogue H-1414.0005BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- Species:HumanBovineMouse
- CAS No.:149234-04-8
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237794
- Protein synonyms:TS1 (1016-1023)
- Protein/peptide name:Thrombospondin-1 (1016-1023)
- Molecular weight:1128.41
- Sequence:H-Arg-Phe-Tyr-Val-Val-Met-Trp-Lys-OH
- Molecular formula:C₅₆H₈₁N₁₃O₁₀S
- Cat. no.:H-1414.0005BA
- Supplier No.:H-1414.0005
Spécifications
A propos de cet article
The octapeptide RFYVVMWK constitutes the active sequence within the 30-mer peptide named C₄ as it is essential for the cell attachment activity of the TS1 CBD. It supports the attachment of G361 melanoma cells and inhibits their adhesion to the rec CBD of TS1. Moreover, this peptide appears to be highly conserved in all TS1 isoforms, and a related sequence is also present in the fragment F9 of laminin. H-1414 (4N1) has been used as CD47 agonist (see also H-6414).
Customers Who Bought This Also Bought