ACTH (1-13)
# de catalogue H-1135.0001BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:22006-64-0
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00133030
- Protein synonyms:Adrenocorticotropic hormone
- Protein/peptide name:ACTH (1-13)
- Molecular weight:1623.86
- Sequence:H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-OH
- Molecular formula:C₇₅H₁₀₆N₂₀O₁₉S
- Cat. no.:H-1135.0001BA
- Supplier No.:H-1135.0001
Spécifications
A propos de cet article
Please see also α-MSH (corresponds to acetyl-ACTH (1-13) amide, H-1075), α-MSH (free acid) (acetyl-ACTH (1-13), H-1070), and (Des-acetyl)-α-MSH (H-4390). Wied: Pituitary Adrenal System Hormones and Behaviour. Symposium on Developments in Endocrinology (1976) / Anon.: ACTH and Related Peptides: Structure, Regulation, and Action. Ann. N.Y. Acad. Sci. 297, 1 (1977) / A.V.Schally: Aspects of Hypothalamic Regulation of the Pituitary Gland. Science 202, 18 (1978) / R.Schwyzer: Studies on Polypeptide Receptors. A Critical View on the Mechanism of ACTH Action. Bull. Schweiz. Acad. Med. Wiss. 34, 263 (1978).
Customers Who Bought This Also Bought