H-Gly-Cys-OH
# de catalogue G-4440.0250BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:57281-78-4
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00672396
- Protein/peptide name:H-Gly-Cys-OH
- Molecular weight:178.21
- Sequence:H-Gly-Cys-OH
- Molecular formula:C₅H₁₀N₂O₃S
- Cat. no.:G-4440.0250BA
- Supplier No.:G-4440.0250
Spécifications
A propos de cet article
Inhibitor of carboxypeptidase U (thrombin activatable fibrinolysis inhibitor, TAFI), Ki 0.14 μM.
Customers Who Bought This Also Bought