H-His-Pro-NH₂ · 2 HBr
Catalog # G-4185.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:59760-04-2
- Size:250 mg
- Protein/peptide name:H-His-Pro-NH₂ · 2 HBr
- MDL No.:MFCD00237991
- Sequence:H-His-Pro-NH₂ · 2 HBr
- Molecular weight:413.11
- Molecular formula:C₁₁H₁₇N₅O₂ · 2HBr
- Storage conditions:–20 ± 5 °C
- Cat. no.:G-4185.0250BA
- Supplier No.:G-4185.0250
Specifications
About this item
The dipeptide His-Pro-amide represents the initial enzymatic degradation product of TRH in plasma. CAS Number (net): 33605-69-5.
Customers Who Bought This Also Bought