H-Arg-Ala-OH Acetate, Bachem
Fournisseur: Bachem Americas
Certificats
A propos de cet article
A salt taste-enhancing dipeptide. Arg-Ala inhibits angiotensin 1-converting enzyme (ACE) and yeast bleomycin hydrolase.
Customers Who Bought This Also Bought