N-2-Cyanoethyl-Val-Leu-anilide
Catalog # G-4285.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:194351-52-5
- Size:50 mg
- Protein/peptide name:N-2-Cyanoethyl-Val-Leu-anilide
- MDL No.:MFCD00237063
- Sequence:N-2-Cyanoethyl-Val-Leu-anilide
- Molecular weight:358.48
- Molecular formula:C₂₀H₃₀N₄O₂
- Storage conditions:–20 ± 5 °C
- Cat. no.:G-4285.0050BA
- Supplier No.:G-4285.0050
Specifications
About this item
These N-modified peptides are used as standards for the determination of the extent of the N-terminal globin modification in erythrocytes of people exposed to alkylating agents or to triazinyl derivatives. Isolated globin is submitted to Edman degradation and the thiohydantoin derivatives are separated and quantified by GC-MS, with the help of these standards.
Customers Who Bought This Also Bought