H-Gly-Gly-OH
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Standard acceptor in γ-glutamyltransferase assays, e.g. with H-Glu(pNA)-OH (L-1265) or H-Glu(AMC)-OH (I-1185).
Customers Who Bought This Also Bought
Standard acceptor in γ-glutamyltransferase assays, e.g. with H-Glu(pNA)-OH (L-1265) or H-Glu(AMC)-OH (I-1185).