H-Ala-Gln-OH, Bachem
Fournisseur: Bachem Americas
Certificats
A propos de cet article
The dipeptides H-Ala-Gln-OH and H-Gly-Gln-OH (N-1070) are stable substitutes for Gln in cell culture media, which tolerate autoclaving and liberate a lower amount of undesired ammonia than Gln. Ala-Gln and Gly-Gln have also found use as Gln sources in parenteral nutrition.
Customers Who Bought This Also Bought