Distillation Receiver with Side Arm, Ace Glass
# de catalogue 89074-450
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- ST Joint No.:14/20 (side arm)
- Description:Side-Arm Distillation Receiver
- Cat. no.:89074-450
- Supplier No.:9373-06
Spécifications
A propos de cet article
The apparatus is graduated to 15mL in 1/10mL subdivisions, with the lower 0.5mL contained in a precision tip for better accuracy with extremely small samples
Fabricated from borosilicate glass.
Customers Who Bought This Also Bought