Electromagnetic Coil, Plunger Principle, Ace Glass
# de catalogue 89055-676
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Electromagnetic Coil
- Cat. no.:89055-676
- Supplier No.:6590-10
Spécifications
A propos de cet article
This coil is designed to operate automatic distilling heads which operate on the plunger principle, whereby the glass rod is pulled up through the center of the magnet.
For operation at 120 volts, 50/60 Hz AC maximum, or at 24 volts DC maximum
For intermittent service only; 30 seconds maximum ON, 25% maximum duty cycle.
Diameter of the hole is 12.7 mm; Diameter of the coil is 38 mm; height is 32 mm.
Ordering information: It is supplied complete with necessary wiring to connect into the system.
Customers Who Bought This Also Bought