Soil Moisture Indicator
# de catalogue 470137-768
Fournisseur: GLOBAL SPECIALTIES SE
Spécifications
- Description:Soil Moisture Indicator
- Cat. no.:470137-768
- Dimensions:1.59×0.84"
- Supplier No.:GSK-908
Spécifications
A propos de cet article
Simple, Accurate Tracking
- Visible indicator
A four LED circuit indicates the amount of moisture in the soil. Higher moisture levels light up more LEDs.
Customers Who Bought This Also Bought