Solar Fan
# de catalogue 470137-764
Fournisseur: GLOBAL SPECIALTIES SE
Spécifications
- Description:Solar Fan
- Cat. no.:470137-764
- Supplier No.:GSK-1001
Spécifications
A propos de cet article
Makes Solar Energy Easy to Understand
Use this kit to create a circuit for converting sunlight into electrical energy. The fan speed is controlled by the intensity of the sunlight.
Customers Who Bought This Also Bought