Classic Wood Standing Height Base Cabinets, Cupboard and Drawer Cabinets, Diversified Woodcrafts
Supplier: VWR International
Certificates
About this item
Classic oak laboratory grade cabinets.
- Classic neutral color
Vertical grain matched select oak veneers finished with a VOC-free, environmentally friendly chemical-resistant finish. Cabinets stained with a light neutral finish. Plywood core cabinet body with removable backs, full extension drawer slides and stainless steel hinges. 96 mm classic wire pulls, twin-pin seismic rated shelf pins, and nylon roller door catches.
Certifications: SEFA Tested.
Ordering information: Units are shipped wrapped and palletized via common carrier trucking. All cabinets include a lifetime warranty on manufacturing parts and processes.
Specifications
- Catalog No:
- 89371-748
- 89372-170
- 89371-916
- 89371-700
- 89371-898
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green