Rodaviss® O-Rings, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Designed for use with Rodaviss® joints and caps to form a leak-tight seal
Available in Nitrile or Viton formats. For threaded joints, see 89059-696 series. For caps, see 89059-718 series.
Customers Who Bought This Also Bought