Four-Neck Reaction Flask Head, Flat Flange, with #7 Ace-Thred, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Designed for use with ground flat flange flasks
With three in-line [ST] joints and an internally threaded #7 Ace-Thred side opening for thermometer (#7 accepts 6–7mm O.D.). Thread forms a compression seal with a nylon bushing and permits vertical adjustment of thermometer. Fabricated from borosilicate glass.
Ordering information: Supplied complete with bushing 89051-122 and FETFE O-Ring 89060-958 (size -008). Uses clamps such as 89055-038, 89055-040, 89055-046, or 89055-048. All clamps must be purchased separately.
Customers Who Bought This Also Bought