Stirring Shaft, PTFE-Coated Stainless Steel, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
PTFE covered stainless steel stirring shafts feature a drilled-type construction and a 10mm O.D
If used un-lubricated, we recommend a 500rpm maximum with a water cooled bearing. A 100rpm maximum is recommended with standard non-water cooled bearings. With condensate film as an added heat transfer agent, 2000rpm can be attained.
BodyTextAccessories: PTFE Gaskets are designed to fit a variety of stirring shafts, such as 89061-938, 89061-946, 89084-572, 89061-962, or 89062-014. Gaskets are also useful as a dust cover, or as a replacement for the gasket supplied with collars (such as 89235-694).
Ordering information: The drilled-type shaft only accepts specific agitators. It is compatible with collet-type bearings. The shaft joins to a stirrer motor via a chuck. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought