Air Sampling Tee, Ace Glass Incorporated
# de catalogue 89059-190
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Reduced Ends Air Sampling Tee, with 25 mm Ace-Thred
- Cat. no.:89059-190
- Overall Length:152 mm
- Supplier No.:7495-27
Spécifications
A propos de cet article
Air sampling tee is fabricated from borosilicate glass.
Sampling tee with threads on all three arms for use with bushing or coupling.
Customers Who Bought This Also Bought