Corning® Centrifuge Bottles, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PP, translucent or PC, clear.
- Clear graduations and textured marking spot
- Specially designed cap with a built-in easy grip feature
- Caps are not interchangeable among different size/material bottles
- Extra-thick walls for durability
- Wide-mouth bottle design for improved liquid handling
Centrifuge bottles for large volume processing in cell harvesting, pelleting and protein purification. Allow researchers to decrease the number of centrifugation cycles, improve the laboratory efficiency and boost productivity. Two cap styles: standard and high performance with silicone O-ring. Leak-free processing at maximum rated speeds (Fill volume should not exceed 80% of the total bottle capacity).
Customers Who Bought This Also Bought