Gooch Crucible, High Form, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
High Form Gooch crucible features an integral fritted disc that is 30mm in diameter
The height above the disc is 45mm. Available in five different porosities. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought