Allihn Tube with Frit, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The Allihn tube features an integral fritted disc with a diameter of 30mm
The height above the disc is 100mm, and the diameter of the bottom tube is 8mm. Available in four different porosities. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought