Single Stack Paper Towel Holder, PETG, TrippNT
Catalogue # 89233-144
Fournisseur: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Paper Towel Holder
- Cat. no.:89233-144
- W×D×H:28 x 10 x 18 cm (11 x 4 x 7")
- Weight:0.5 kg (1 lbs.)
- Supplier No.:50709
Spécifications
À propos de cet article
Clear PETG single stack paper towel holder has a simple design that allows quick access to one or several C-fold, Z-fold, Tri-fold or multi-fold paper towels
Made from extra-tough PETG which won't break like acrylic and is clear for easy viewing. Holds about 150 folded paper towels and can sit on the counter or be mounted on the wall with included tape or keyholes.