Platinum Dishes; Standard and Evaporation Shapes, XRF Scientific
Fournisseur: Kitco Metals
Certificats
A propos de cet article
These 99.95% pure platinum dishes are available in standard and evaporation forms.
- High temperature strength
- Ductility; corrosion; and oxidation resistance
- Excellent resistance to chemical attacks by acids and fusion mixtures
Due to their high temperature strength; ductility; corrosion and oxidation resistance; platinum dishes are critical when high temperatures and resistance to chemical attacks are required.
Ideally suited for fusions; wet chemistry; digestion; ash testing; and Loss on Ignition (LOI) applications.
Certifications: The 100 ml dishes; in both standard and evaporation forms; meets ASTM Standard IP 501.
Ordering information: Dish lids are sold separately.
Spécifications
- Catalog No:
- 89401-336
- 89401-314
- 89401-312
- 89401-334
- 89401-310
- 89401-332
- 89401-308
- 89401-330
- 89401-306
- 89401-328
- 89401-304
- 89401-326
- 89401-324
- 89401-322
- 89401-320
- 89401-318
- 89401-316
- Material:
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum
- Platinum