MetalGuard® Heavy Weight 1880 Cut Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
MetalGuard® heavy weight gloves form to hands for comfortability.
- Highest level of cut protection within the MetalGuard® line
- Continuous knit conforms to the hand
MetalGuard® heavy weight gloves ensuring a close, secure fit. Additional reinforcement in the thumb crotch area increases wear life. Gloves are prewashed and preshrunk for a comfortable fit.
Certifications: ANSI Cut Level 5, A6.
Ordering information: These gloves are sold by the dozen pair.
Caution: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades
Specifications
- Catalog No:
- 80091-414
- 80091-418
- 80091-416
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 7
- 7
- 7