A propos de cet article
Whizard® stainless steel gloves with 7¹/₂," cuff are ergonomically designed to be worn on both hands.
Patented self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten.
- Ergonomically designed
- Patented self-adjusting tensioning system
- Micro plasma welded rings
- No gaps to trap soil
- Ambidextrous
- Easy to clean
Whizard® stainless steel gloves with 7¹/₂," cuff are ergonomically designed to be worn on both hands.
Whizard® stainless steel gloves provide superior cut and puncture protection. They feature a patented self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten. All Whizard® mesh gloves come with a metal serial number tag, which helps with reissuance and sizing information. Micro plasma welded rings have no gaps to trap soil that can feed bacterial growth. These gloves are made with 4 mm ring using 0.5 mm wire. Gloves are ambidextrous; just turn inside out for the other hand. These gloves are easy to clean.
Whizard® stainless steel gloves provide superior cut and puncture protection. They feature a patented self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten. All Whizard® mesh gloves come with a metal serial number tag, which helps with reissuance and sizing information. Micro plasma welded rings have no gaps to trap soil that can feed bacterial growth. These gloves are made with 4 mm ring using 0.5 mm wire. Gloves are ambidextrous; just turn inside out for the other hand. These gloves are easy to clean.
Spécifications
- Catalog No:
- 80091-086
- 80091-084
- 80091-082
- 80091-080
- 80091-078
- 80091-076
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff length:
- 7 1/2"
- 7 1/2"
- 7 1/2"
- 7 1/2"
- 7 1/2"
- 7 1/2"